1-(Trimethylsilyl)-1,4-pentadiyne ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Trimethylsilyl-1,4-pentadiyne
H53436.03 H53436.06 H53436.14
H53436.03EA 995 SEK
H53436.03 H53436.06 H53436.14
1-(Trimethylsilyl)-1,4-pentadiyne ≥98%
1-(Trimethylsilyl)-1,4-pentadiyne
Purity: 98% CAS No.: 71789-10-1 Molecular Formula: C8H12Si Formula Weight: 136.27 Flash Point: 8°(46°F) Boiling Point: 42°/25mm UN No.: 1993 MDL No.: MFCD00066350
Formel: C₈H₁₂Si
Kokpunkt: 42 °C (25 mmHg)
Densitet: 0,789
Flampunkt: 8 °C (46 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00066350
CAS nummer: 71789-10-1
ADR: 3,II