3,5-Difluorophenylhydrazine hydrochloride ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: (3,5-Difluorophenyl)hydrazine hydrochloride
H26189.03 H26189.06
H26189.03EA 935 SEK
H26189.03 H26189.06
3,5-Difluorophenylhydrazine hydrochloride ≥97%
3,5-Difluorophenylhydrazine hydrochloride
Purity: 97% CAS No.: 134993-88-7 Molecular Formula: C6H6F2N2.HCl Formula Weight: 180.59 Melting Point: 264-266° MDL No.: MFCD03094171
Formel: C₆H₇Cl₃N₂
MW: 213,49 g/mol
Smältpunkt: 264…266 °C
Storage Temperature: Omgivande
MDL Number: MFCD03094171
CAS nummer: 134993-88-7