2-(Trifluoromethyl)phenylhydrazinium chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(Trifluoromethyl)phenylhydrazine hydrochloride, 1-(2-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 2-Hydrazinobenzotrifluoride hydrochloride, (2-(Trifluormethyl)phenyl)hydrazin HCl
B25182.06 B25182.14
B25182.06EA 1625 SEK
B25182.06 B25182.14
2-(Trifluoromethyl)phenylhydrazinium chloride ≥98%
2-(Trifluoromethyl)phenylhydrazinium chloride
Purity: 98% CAS No.: 3107-34-4 Molecular Formula: C7H7F3N2.HCl Formula Weight: 212.60 Melting Point: ca 235° dec. Beilstein Reference: 4582303
Formel: C₇H₈ClF₃N₂
MW: 212,6 g/mol
Smältpunkt: ∼ 235 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00102619
CAS nummer: 3107-34-4