Ethyl pelargonate ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl nonanoate, Pelargonic acid ethyl ester, Nonanoic acid ethyl ester
B24190.AE B24190.AP
B24190.AEEA 780 SEK
B24190.AE B24190.AP
Ethyl pelargonate ≥97%
Ethyl pelargonate
Purity: 97% CAS No.: 123-29-5 Molecular Formula: C11H22O2 Formula Weight: 186.30 Flash Point: 94°(201°F) Refractive Index: 1.4220 Odour: Pleasant Melting Point: -44° Boiling Point: 227-229° Beilstein Reference: 1759169 Merck Reference: 14,3838 MDL No.: MFCD00009570
Formel: C₁₁H₂₂O₂
Kokpunkt: 227…229 °C
Smältpunkt: –44 °C
Densitet: 0,866
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009570
CAS nummer: 123-29-5
EINECS: 204-615-7
Merck Index: 14,3838