Allylchlorodimethylsilane 95%, rods

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Allyldimethylchlorosilane, ADMCS

B22237.06 B22237.09 B22237.14
B22237.06EA 1475 SEK
B22237.06 B22237.09 B22237.14
Allylchlorodimethylsilane 95%, rods
Allylchlorodimethylsilane

Purity: 97% CAS No.: 4028-23-3 Molecular Formula: CH2=CHCH2Si(CH3)2Cl Formula Weight: 134.68 Flash Point: 5°(41°F) Refractive Index: 1.4290 Boiling Point: 110-112° Beilstein Reference: 1737978 UN No.: 2985 MDL No.: MFCD00000506

Formel: C₅H₁₁ClSi
MW: 134.68 g/mol
Kokpunkt: 110…112 °C
Densitet: 0.896
Flampunkt: 5 °C (41 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000506
CAS nummer: 4028-23-3
EINECS: 223-706-2
UN: 2985
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR