2,4-Diamino-6-phenyl-1,3,5-triazine 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Benzoguanamine, 2,4-Diamino-6-phenyl-s-triazine, 6-Phenyl-1,3,5-triazine-2,4-diyldiamine, 6-Phenyl-1,3,5-triazine-2,4-diamine, 2,6-Diamino-4-phenyl-1,3,5-triazine
B20157.0E B20157.22 B20157.36
B20157.0EEA 5100 SEK
B20157.0E B20157.22 B20157.36
2,4-Diamino-6-phenyl-1,3,5-triazine 99%
2,4-Diamino-6-phenyl-1,3,5-triazine
Purity: 99% CAS No.: 91-76-9 Molecular Formula: C9H9N5 Formula Weight: 187.21 Melting Point: 225-228° Beilstein Reference: 153223 Merck Reference: 14,1089 MDL No.: MFCD00023187
Formel: C₉H₉N₅
MW: 187,2 g/mol
Smältpunkt: 225…228 °C
Storage Temperature: Omgivande
MDL Number: MFCD00023187
CAS nummer: 91-76-9
EINECS: 202-095-6
Merck Index: 14,1089