Diisopropanolamin sum of isomers ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A18451.0B A18451.30
A18451.0BEA 1425 SEK
A18451.0B A18451.30
Diisopropanolamin sum of isomers ≥98%
Diisopropanolamin
Purity: 98+% CAS No.: 110-97-4 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 130°(266°F) Odour: Amine-like Boiling Point: 249-250° Beilstein Reference: 605363 Merck Reference: 14,3195 MDL No.: MFCD00004531
Formel: C₆H₁₅NO₂
Kokpunkt: 249…250 °C
Smältpunkt: 38…45 °C
Densitet: 1,004
Flampunkt: 130 °C (266 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004531
CAS nummer: 110-97-4
EINECS: 203-820-9
Merck Index: 14,3195