Sodium oxamate ≥98%
Leverantör: Thermo Scientific
Synonyms:
Oxamic acid sodium salt, Sodium 2-amino-2-oxoacetate
A16532.06
A16532.14
A16532.06EA
1200
SEK
A16532.06
A16532.14
Sodium oxamate ≥98%
Sodium oxamate
Purity: 98%
CAS No.: 565-73-1
Molecular Formula: H2NC(O)CO2Na
Formula Weight: 111.03
Melting Point:
Formel:
C₂H₂NNaO₃ MW: 111,03 g/mol Smältpunkt: >300 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00044553 CAS nummer: 565-73-1 EINECS: 209-290-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...