Methyl-2-bromo-5-methoxybenzoate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A15322.06 A15322.14
A15322.06EA 1325 SEK
A15322.06 A15322.14
Methyl-2-bromo-5-methoxybenzoate ≥98%
Methyl-2-bromo-5-methoxybenzoate
Purity: 98% CAS No.: 35450-36-3 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Flash Point: 110°(230°F) Refractive Index: 1.5640 Boiling Point: 271° Beilstein Reference: 2641378 MDL No.: MFCD00051594
Formel: C₉H₉BrO₃
Kokpunkt: 271 °C
Densitet: 1,52
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051594
CAS nummer: 35450-36-3