3,5-Dichlorophenylhydrazinium chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride
A15127.06 A15127.14
A15127.06EA 2350 SEK
A15127.06 A15127.14
3,5-Dichlorophenylhydrazinium chloride ≥97%
3,5-Dichlorophenylhydrazinium chloride
Purity: 97% CAS No.: 63352-99-8 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.50 Melting Point: ca 204° dec. Beilstein Reference: 4208459
Formel: C₆H₇Cl₃N₂
MW: 213,49 g/mol
Smältpunkt: ∼ 204 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012938
CAS nummer: 63352-99-8
EINECS: 264-109-7