(Chloromethyl)dimethylchlorosilane ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: CMDMCS, Chloro(chloromethyl)dimethylsilane
A14899.09 A14899.14 A14899.22
A14899.09EA 850 SEK
A14899.09 A14899.14 A14899.22
(Chloromethyl)dimethylchlorosilane ≥98%
(Chloromethyl)dimethylchlorosilane
Purity: 98% CAS No.: 1719-57-9 Molecular Formula: C3H8Cl2Si Formula Weight: 143.09 Flash Point: 21°(69°F) Refractive Index: 1.4370 Odour: Acrid Boiling Point: 110-113° Beilstein Reference: 1731933 UN No.: 2985 MDL No.: MFCD00000875
Formel: C₃H₈Cl₂Si
MW: 143,09 g/mol
Kokpunkt: 110…113 °C
Densitet: 1,082
Flampunkt: 21 °C (69 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000875
CAS nummer: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II