2-(Trifluoromethyl)phenylhydrazine ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine, 2-Hydrazinobenzotrifluoride
A14781.03 A14781.06
A14781.03EA 975 SEK
A14781.03 A14781.06
2-(Trifluoromethyl)phenylhydrazine ≥97%
2-(Trifluoromethyl)phenylhydrazine
Purity: 97% CAS No.: 365-34-4 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Melting Point: 57-63° Beilstein Reference: 641592
Formel: C₇H₇F₃N₂
MW: 176,14 g/mol
Smältpunkt: 57…63 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052687
CAS nummer: 365-34-4