2-Chlorophenylhydrazinium chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Chlorophenylhydrazine hydrochloride, (2-Chlorophenyl)hydrazine hydrochloride, 1-(2-Chlorophenyl)hydrazine hydrochloride
A14136.06 A14136.14 A14136.22
A14136.06EA 745 SEK
A14136.06 A14136.14 A14136.22
2-Chlorophenylhydrazinium chloride ≥97%
2-Chlorophenylhydrazinium chloride
Purity: 97% CAS No.: 41052-75-9 Molecular Formula: C6H7ClN2.HCl Formula Weight: 179.05 Melting Point: ca 201° dec. Beilstein Reference: 3699381
Formel: C₆H₈Cl₂N₂
MW: 179,05 g/mol
Smältpunkt: ∼ 201 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012928
CAS nummer: 41052-75-9
EINECS: 255-194-1