3,4-Dichlorophenylhydrazinium chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,4-Dichlorophenylhydrazine hydrochloride, 1-(3,4-Dichlorophenyl)hydrazine hydrochloride
A13906.09 A13906.18 A13906.30
A13906.09EA 1600 SEK
A13906.09 A13906.18 A13906.30
3,4-Dichlorophenylhydrazinium chloride ≥98%
3,4-Dichlorophenylhydrazinium chloride
Purity: 98+% CAS No.: 19763-90-7 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.50 Melting Point: ca 229° dec. Beilstein Reference: 3703934
Formel: C₆H₇Cl₃N₂
MW: 213,49 g/mol
Smältpunkt: ∼ 229 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012937
CAS nummer: 19763-90-7
EINECS: 243-278-0