Benzophenone hydrazone ≥98%

Leverantör: Thermo Scientific
Fara

A12266.0B A12266.18 A12266.30
A12266.0BEA 5725 SEK
A12266.0B A12266.18 A12266.30
Benzophenone hydrazone ≥98%
Benzophenone hydrazone

Purity: 98+% CAS No.: 5350-57-2 Molecular Formula: C13H12N2 Formula Weight: 196.25 Flash Point: 146°(294°F) Odour: Characteristic Boiling Point: 225-230°/55mm Beilstein Reference: 1910177 UN No.: 2811 MDL No.: MFCD00007624

Formel: C₁₃H₁₂N₂
MW: 196,25 g/mol
Kokpunkt: 225…230 °C (55 mmHg)
Smältpunkt: 96…100 °C
Densitet: 1,1
Flampunkt: 146 °C (294 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007624
CAS nummer: 5350-57-2
EINECS: 226-321-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR