4-Bromophenyl methyl sulphone ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-4-(methylsulphonyl)benzene
A11427.09 A11427.18
A11427.09EA 1150 SEK
A11427.09 A11427.18
4-Bromophenyl methyl sulphone ≥98%
4-Bromophenyl methyl sulphone
Purity: 98+% CAS No.: 3466-32-8 Molecular Formula: C7H7BrO2S Formula Weight: 235.10 Melting Point: 101-102° Beilstein Reference: 2045675
Formel: C₇H₇BrO₂S
MW: 235.1 g/mol
Smältpunkt: 101…102 °C
Storage Temperature: Omgivande
MDL Number: MFCD00025065
CAS nummer: 3466-32-8
EINECS: 222-421-0