(±)-6-Methyl-5-hepten-2-ol 99%
Leverantör: Thermo Fisher Scientific
Synonyms: (±)-2-Methyl-2-hepten-6-ol, (±)-2-Methylhept-2-en-6-ol, (±)-6-Methylhept-5-en-2-ol
187690250 187691000
ACRO187690250EA 1725 SEK
ACRO187690250 ACRO187691000
(±)-6-Methyl-5-hepten-2-ol 99%
(±)-6-Methyl-5-hepten-2-ol
Formel: C₈H₁₆O
Kokpunkt: 78 °C (14 mmHg)
Densitet: 0,84 g/cm³
Flampunkt: 67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004561
CAS nummer: 4630-06-2
EINECS: 216-377-1
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4475 to 1.4495 (20°C, 589 nm)
GC ≥98.5 %