1-Bromo-4-fluorobenzene ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Bromofluorobenzene
A11630.0E A11630.22 A11630.36
A11630.0EEA 11300 SEK
A11630.0E A11630.22 A11630.36
1-Bromo-4-fluorobenzene ≥99%
1-Bromo-4-fluorobenzene
Purity: 99% CAS No.: 460-00-4 Molecular Formula: C6H4BrF Formula Weight: 175.01 Flash Point: 60°(140°F) Refractive Index: 1.5270 Odour: Aromatic Melting Point: -16° Boiling Point: 151-153° Beilstein Reference: 774102 UN No.: 1993 MDL No.: MFCD00000342
Formel: C₆H₄BrF
Kokpunkt: 151…153 °C
Smältpunkt: –16 °C
Densitet: 1,604
Flampunkt: 60 °C (140 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000342
CAS nummer: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3,III