Sulfolan 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: Tetrahydrotiofen-1,1-dioxid
138432500 138430010 138430025 138430100
ACRO138432500EA 1300 SEK
ACRO138432500 ACRO138430010 ACRO138430025 ACRO138430100
Sulfolan 99+%
Sulfolan
Formel: C₄H₈O₂S
MW: 120,17 g/mol
Kokpunkt: 285 °C (760 mmHg)
Smältpunkt: 27,4…27.8 °C
Densitet: 1,26 g/cm³
Flampunkt: 165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005484
CAS nummer: 126-33-0
EINECS: 204-783-1
UN: 3335
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4820 to 1.4850 (20°C, 589 nm)
Water ≤0.3 %
Sulfated ash ≤0.1 %
GC ≥99.0 %