1-Brom-3,5-difluorbenzen ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Difluorobromobenzene
A14053.06 A14053.09 A14053.14 A14053.22 A14053.30
A14053.06EA 730 SEK
A14053.06 A14053.09 A14053.14 A14053.22 A14053.30
1-Brom-3,5-difluorbenzen ≥98%
1-Brom-3,5-difluorbenzen
Purity: 98% CAS No.: 461-96-1 Molecular Formula: C6H3BrF2 Formula Weight: 192.99 Flash Point: 44°(111°F) Refractive Index: 1.4990 Melting Point: -27° Boiling Point: 140-141° Beilstein Reference: 1931520 UN No.: 1993 MDL No.: MFCD00010305
Formel: C₆H₃BrF₂
MW: 192,99 g/mol
Kokpunkt: 140…141 °C
Smältpunkt: –27 °C
Densitet: 1,676
Flampunkt: 44 °C (111 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010305
CAS nummer: 461-96-1
EINECS: 416-710-2
UN: 1993
ADR: 3,III