4-Brom-1,2-difluorbenzen ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Brom-3,4-difluorbenzen
A11934.06 A11934.14 A11934.22
A11934.06EA 685 SEK
A11934.06 A11934.14 A11934.22
4-Brom-1,2-difluorbenzen ≥98%
4-Brom-1,2-difluorbenzen
Purity: 98+% CAS No.: 348-61-8 Molecular Formula: C6H3BrF2 Formula Weight: 192.99 Flash Point: 33°(91°F) Refractive Index: 1.5050 Melting Point: -4° Boiling Point: 150-151° Beilstein Reference: 1934811 UN No.: 1993 MDL No.: MFCD00000304
Formel: C₆H₃BrF₂
MW: 192,99 g/mol
Kokpunkt: 150…151 °C
Smältpunkt: –4 °C
Densitet: 1,707
Flampunkt: 33 °C (91 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000304
CAS nummer: 348-61-8
EINECS: 206-481-5
UN: 1993
ADR: 3,III