1-Bromo-2-fluorobenzene 99
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Bromofluorobenzene
A10635.14 A10635.22 A10635.36
A10635.14EA 1025 SEK
A10635.14 A10635.22 A10635.36
1-Bromo-2-fluorobenzene 99
1-Bromo-2-fluorobenzene
Purity: 99% CAS No.: 1072-85-1 Molecular Formula: C6H4BrF Formula Weight: 175.01 Flash Point: 43°(109°F) Refractive Index: 1.5340 Odour: Aromatic Melting Point: -8° Boiling Point: 155-157° Beilstein Reference: 1071819 UN No.: 1993 MDL No.: MFCD00000282
Formel: C₆H₄BrF
MW: 175 g/mol
Kokpunkt: 155…157 °C
Smältpunkt: –8 °C
Densitet: 1.601
Flampunkt: 43 °C (109 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000282
CAS nummer: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3 III
Order Now
SPECIFIKATIONER
Environmentally Preferable