Flaskor, smal hals, graderad, fyrkantig, PPCO, med skruvlock, Nalgene®
Leverantör: Thermo Fisher Scientific
312016-0060 312016-0125 312016-0500 312016-1000
NALG312016-0060EA 25675 SEK
NALG312016-0060 NALG312016-0125 NALG312016-0500 NALG312016-1000
Flaskor, smal hals, graderad, fyrkantig, PPCO, med skruvlock, Nalgene®
Flaskor Flaskor, plast
PPCO, translucent, with PP screw cap.
  • Excellent chemical and temperature resistance
  • Natural, polypropylene screw closure
  • Drip-proof neck finish for convenient dispensing
  • Graduated, autoclavable and leakproof
  • Resistant to acids, alcohols and bases
  • Temperature range of 0 to 135°C
Easily measure contents with molded-in graduations on the Nalgene™ square natural PPCO graduated bottles with closure. These bottles provide visible liquid levels and feature a space-saving square design.