4-Bromo-2-chlorophenol 99%
Leverantör: Thermo Fisher Scientific
Fara
173530250 173531000 173530050
ACRO173530250EA 1450 SEK
ACRO173530250 ACRO173531000 ACRO173530050
4-Bromo-2-chlorophenol 99%
4-Bromo-2-chlorophenol
Formel: C₆H₄BrClO
MW: 207.45 g/mol
Kokpunkt: 232
Smältpunkt: 47…51 °C
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002166
CAS nummer: 3964-56-5
EINECS: 223-572-5
UN: 2811
ADR: 6.1 II
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or needles and/or chunks or low melting mass
Infrared spectrum Conforms
Melting point 47 to 51 °C
GC ≥98.5 %