1-Bromo-2-chloro-4-nitrobenzene ≥98%
Leverantör: Thermo Scientific
Fara
L07573.09 L07573.18
L07573.09EA 1475 SEK
L07573.09 L07573.18
1-Bromo-2-chloro-4-nitrobenzene ≥98%
1-Bromo-2-chloro-4-nitrobenzene
Purity: 98% CAS No.: 29682-39-1 Molecular Formula: C6H3BrClNO2 Formula Weight: 236.45 Melting Point: 59-62° Beilstein Reference: 2253107 UN No.: 2811 MDL No.: MFCD00051514
Formel: C₆H₃BrClNO₂
Smältpunkt: 59…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051514
CAS nummer: 29682-39-1
EINECS: 249-773-8
UN: 2811
ADR: 6.1,III