4-Bromo-2-chloroaniline ≥98%
Leverantör: Thermo Scientific
Fara
A17859.09 A17859.18 A17859.30
A17859.09EA 1100 SEK
A17859.09 A17859.18 A17859.30
4-Bromo-2-chloroaniline ≥98%
4-Bromo-2-chloroaniline
Purity: 98+% CAS No.: 38762-41-3 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Melting Point: 68-72° Beilstein Reference: 3117740
Formel: C₆H₅BrClN
MW: 206,47 g/mol
Kokpunkt: 264 °C (1013 hPa)
Smältpunkt: 68…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007660
CAS nummer: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: 6.1,III