4-Bromo-2-chloroaniline 99%
Leverantör: Thermo Scientific
Varning
167480100 167480500
ACRO167480100EA 810 SEK
ACRO167480100 ACRO167480500
4-Bromo-2-chloroaniline 99%
4-Bromo-2-chloroaniline
Formel: C₆H₅BrClN
MW: 206,47 g/mol
Kokpunkt: 264 °C (1013 hPa)
Smältpunkt: 67…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007660
CAS nummer: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥98.5 %