2-Bromo-5-klorofluorobenzen 98
Leverantör: Thermo Fisher Scientific
Varning
A19570.06 A19570.14
A19570.06EA 500 SEK
A19570.06 A19570.14
2-Bromo-5-klorofluorobenzen 98
2-Bromo-5-klorofluorobenzen
Purity: 98% CAS No.: 1996-29-8 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Flash Point: 92°(197°F) Refractive Index: 1.5540 Boiling Point: 70-72°/11mm Beilstein Reference: 2081083 MDL No.: MFCD00079708
Formel: C₆H₃BrClF
MW: 209.44 g/mol
Kokpunkt: 70…72 °C (11 mmHg)
Densitet: 1.678
Flampunkt: 92 °C (197 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00079708
CAS nummer: 1996-29-8