1-Bromo-3-chlorobenzene 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Bromochlorobenzene
154200250 154201000
ACRO154200250EA 490 SEK
ACRO154200250 ACRO154201000
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene
Formel: C₆H₄BrCl
MW: 191,45 g/mol
Kokpunkt: 196 °C (1013 hPa)
Smältpunkt: –21,5 °C
Densitet: 1,63 g/cm³
Flampunkt: 80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000568
CAS nummer: 108-37-2
EINECS: 203-575-8
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5780 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure