5-Bromoanthranilic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Amino-5-bromobenzoic acid
305620050 305620250
ACRO305620050EA 830 SEK
ACRO305620050 ACRO305620250
5-Bromoanthranilic acid 97%
5-Bromoanthranilic acid
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
Kokpunkt: 342 °C (1013 hPa)
Smältpunkt: 218…220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007823
CAS nummer: 5794-88-7
EINECS: 227-338-3
ADR: 6.1,III
Merck Index: 13,01389
Order Now
Specifikation testresultat
Appearance Beige Powder
Infrared spectrum Conforms
GC ≥96.0 %