3,5-Dibromoanthranilic acid

Leverantör: Thermo Scientific
Varning

Synonyms: 2-Amino-3,5-dibromobenzoicacid, 2-Amino-3,5-dibromobenzoic acid

BTB01924EA
MAYBBTB01924EAEA 0 SEK
MAYBBTB01924EA
3,5-Dibromoanthranilic acid
3,5-Dibromoanthranilic acid
Formel: C₇H₅Br₂NO₂
MW: 294,93 g/mol
MDL Number: MFCD00017087
CAS nummer: 609-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR