3-Bromobenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: m-Bromobenzoic Acid
A12606.14 A12606.22
A12606.14EA 945 SEK
A12606.14 A12606.22
3-Bromobenzoic acid ≥98%
3-Bromobenzoic acid
Purity: 98+% CAS No.: 585-76-2 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Melting Point: 153-157° Beilstein Reference: 2041792
Formel: C₇H₅BrO₂
Kokpunkt: 308 °C (1013 hPa)
Smältpunkt: 153…157 °C
Densitet: 1,948
Storage Temperature: Omgivande
MDL Number: MFCD00002487
CAS nummer: 585-76-2
EINECS: 209-562-3