2-Bromobenzoic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: o-Bromobenzoic Acid
160080250 160081000
ACRO160080250EA 595 SEK
ACRO160080250 ACRO160081000
2-Bromobenzoic acid 97%
2-Bromobenzoic acid
Formel: C₇H₅BrO₂
Kokpunkt: 296 °C (1013 hPa)
Smältpunkt: 146…151 °C
Densitet: 1,929 g/cm³ (20 °C)
Flampunkt: ≥ 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002402
CAS nummer: 88-65-3
EINECS: 201-848-6
Order Now
Specifikation testresultat
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 145 to 150 °C
HPLC ≥96.0 %
Fines ≤8.5 % (< 0.5 cm)