Burette stopcocks, straight
Leverantör: Brand
81937 81939
BRND81937EA 850 SEK
BRND81937 BRND81939
Burette stopcocks, straight
Kranar
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.