3,5-Dibromosalicylaldehyde 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde
153400250 153401000
ACRO153400250EA 2300 SEK
ACRO153400250 ACRO153401000
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde
Formel: C₇H₄Br₂O₂
MW: 279,92 g/mol
Kokpunkt: 261 °C (1013 hPa)
Smältpunkt: 80…84 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003318
CAS nummer: 90-59-5
EINECS: 202-003-4
Merck Index: 13,03052
Order Now
Specifikation testresultat
Appearance Yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
Titration with NaOH ≥97.5 %