3-Bromobenzaldehyde 96%
Leverantör: Thermo Fisher Scientific
Varning
106660250 106661000
ACRO106660250EA 660 SEK
ACRO106660250 ACRO106661000
3-Bromobenzaldehyde 96%
3-Bromobenzaldehyde
Formel: C₇H₅BrO
Kokpunkt: 229 °C (1013 hPa)
Smältpunkt: 18…21 °C
Densitet: 1,58 g/cm³
Flampunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003345
CAS nummer: 3132-99-8
EINECS: 221-526-9
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5920 to 1.5960 (20°C, 589 nm)
GC ≥95 %