1,3,5-Tribromobenzene 98%
Leverantör: Thermo Scientific
Varning
161080250 161081000
ACRO161080250EA 1025 SEK
ACRO161080250 ACRO161081000
1,3,5-Tribromobenzene 98%
1,3,5-Tribromobenzene
Formel: C₆H₃Br₃
MW: 314,8 g/mol
Kokpunkt: 271 °C (1013 hPa)
Smältpunkt: 118…123 °C
Densitet: 2,637 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000080
CAS nummer: 626-39-1
EINECS: 210-947-3
Order Now
Specifikation testresultat
Appearance White to orange-brown Powder
Infrared spectrum Conforms
Melting point 118 to 123 °C
GC ≥97.5 %
Color scale ≤100 APHA