9,10-Diphenylanthracene ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A15714.03 A15714.06 A15714.14
A15714.03EA 1600 SEK
A15714.03 A15714.06 A15714.14
9,10-Diphenylanthracene ≥99%
9,10-Diphenylanthracene
Purity: 99% CAS No.: 1499-10-1 Molecular Formula: C26H18 Formula Weight: 330.43 Odour: Odourless Melting Point: 248-254° Beilstein Reference: 1914010 MDL No.: MFCD00001253
Formel: C₂₆H₁₈
Kokpunkt: 485 °C (1013 hPa)
Smältpunkt: 248…254 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001253
CAS nummer: 1499-10-1
EINECS: 216-105-1