Tiron
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt, Catechol-3,5-disulphonic acid disodium salt, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt, 3,5-Pyrocatecholdisulphonic acid disodium salt, Disodium 4,5-dihydroxybenzene-1,3-disulphonate
174140250 174141000 174145000
ACRO174140250EA 1025 SEK
ACRO174140250 ACRO174141000 ACRO174145000
Tiron
Tiron
Formel: C₆H₄Na₂O₈S₂
MW: 314,20 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149531
CAS nummer: 149-45-1
EINECS: 205-741-5
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Water 4.5 to 6.0 % (K.F.)
Titration with NaOH ≥98.0 %
HPLC ≥97.5 %