9-Bromoanthracene 96%

Leverantör: Thermo Scientific
Varning

106640050 106640250
ACRO106640050EA 890 SEK
ACRO106640050 ACRO106640250
9-Bromoanthracene 96%
9-Bromoanthracene
Formel: C₁₄H₉Br
MW: 257,13 g/mol
Kokpunkt: 359 °C (1013 hPa)
Smältpunkt: 98…100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001243
CAS nummer: 1564-64-3
EINECS: 216-359-3

Order Now

Specifikation testresultat

Appearance Yellow to green to brown Powder or crystals or needles
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR