3-Hexyne ≥99%
Leverantör: Thermo Fisher Scientific
Fara
L02682.06 L02682.14
L02682.06EA 780 SEK
L02682.06 L02682.14
3-Hexyne ≥99%
3-Hexyne
Purity: 99% CAS No.: 928-49-4 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -10°(14°F) Refractive Index: 1.4110 Odour: Characteristic Boiling Point: 80-82° Beilstein Reference: 1731158 UN No.: 3295 MDL No.: MFCD00009381
Formel: C₆H₁₀
Kokpunkt: 80…82 °C
Densitet: 0,721
Flampunkt: –10 °C (14 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009381
CAS nummer: 928-49-4
EINECS: 213-173-4
UN: 3295
ADR: 3,II