1-Undecyne 97
Leverantör: Thermo Fisher Scientific
Varning
L02883.09 L02883.18 L02883.04
L02883.09EA 3900 SEK
L02883.09 L02883.18 L02883.04
1-Undecyne 97
1-Undecyne
Purity: 97% CAS No.: 2243-98-3 Molecular Formula: C11H20 Formula Weight: 152.28 Flash Point: 65°(149°F) Refractive Index: 1.4310 Odour: Characteristic Melting Point: -25° Boiling Point: 196° Beilstein Reference: 1740043 MDL No.: MFCD00015063
Formel: C₁₁H₂₀
MW: 152.28 g/mol
Kokpunkt: 196 °C
Smältpunkt: –25 °C
Densitet: 0.770
Flampunkt: 65 °C (149 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00015063
CAS nummer: 2243-98-3
EINECS: 218-825-1