1-Hexyne ≥98%
Leverantör: Thermo Scientific
Fara
A13156.09 A13156.18 A13156.30
A13156.09EA 555 SEK
A13156.09 A13156.18 A13156.30
1-Hexyne ≥98%
1-Hexyne
Purity: 98+% CAS No.: 693-02-7 Molecular Formula: CH3(CH2)3C≡CH Formula Weight: 82.15 Flash Point: -21°(-5°F) Refractive Index: 1.3990 Odour: Characteristic Melting Point: -132° Boiling Point: 69-71° Beilstein Reference: 635687 UN No.: 3295 MDL No.: MFCD00009504
Formel: C₆H₁₀
MW: 82,15 g/mol
Kokpunkt: 69…71 °C
Smältpunkt: –132 °C
Densitet: 0,718
Flampunkt: –21 °C (–5 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009504
CAS nummer: 693-02-7
EINECS: 211-736-9
UN: 3295
ADR: 3,II