1-Hexadecyne tech. 90%, Technical Grade
Leverantör: Thermo Scientific
A11366.06
A11366.14
A11366.06EA
4475
SEK
A11366.06
A11366.14
1-Hexadecyne tech. 90%, Technical Grade
1-Hexadecyne
Purity: 90%
CAS No.: 629-74-3
Molecular Formula: C16H30
Formula Weight: 222.42
Flash Point:
Formel:
C₁₆H₃₀ Kokpunkt: 146…148 °C (10 mmHg) Smältpunkt: 13…16 °C Densitet: 0,790 Flampunkt: >110 °C (230 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00015084 CAS nummer: 629-74-3 EINECS: 211-106-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...