1-Dodecyne 98%
Leverantör: Thermo Fisher Scientific
Varning
223450050 223450250
ACRO223450050EA 1025 SEK
ACRO223450050 ACRO223450250
1-Dodecyne 98%
1-Dodecyne
Formel: C₁₂H₂₂
Kokpunkt: 215 °C (760 mmHg)
Smältpunkt: –19 °C
Densitet: 0,77 g/cm³
Flampunkt: 79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008960
CAS nummer: 765-03-7
EINECS: 212-134-9
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥97.5 %