1,1-Bis(methylthio)-2-nitroethylene ≥99%
Leverantör: Thermo Scientific
Varning
A11789.06 A11789.14 A11789.22
A11789.06EA 1100 SEK
A11789.06 A11789.14 A11789.22
1,1-Bis(methylthio)-2-nitroethylene ≥99%
1,1-Bis(methylthio)-2-nitroethylene
Purity: 99% CAS No.: 13623-94-4 Molecular Formula: C4H7NO2S2 Formula Weight: 165.23 Odour: Unpleasant Melting Point: 124-127° Beilstein Reference: 1524538 MDL No.: MFCD00010443
Formel: C₄H₇NO₂S₂
MW: 165,24 g/mol
Smältpunkt: 124…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010443
CAS nummer: 13623-94-4
EINECS: 237-108-4
UN: 2811
ADR: 6.1,II