Tetrakloreten, EMPLURA®, Supelco®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ200
1.00964.1000 1.00964.2500 1.00964.6025 1.00964.9190
1.00964.1000EA 1650 SEK
1.00964.1000 1.00964.2500 1.00964.6025 1.00964.9190
Tetrakloreten, EMPLURA®, Supelco®
Tetrakloreten
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel: C₂Cl₄
Kokpunkt: 121,1 °C (1013 hPa)
Smältpunkt: –22 °C
Densitet: 1,63 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000834
CAS nummer: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
Merck Index: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM