trans-1,4-Dibromo-2-butene 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene
406460250 406461000 406465000
ACRO406460250EA 1525 SEK
ACRO406460250 ACRO406461000 ACRO406465000
trans-1,4-Dibromo-2-butene 97%
trans-1,4-Dibromo-2-butene
Formel: C₄H₆Br₂
MW: 213.9 g/mol
Kokpunkt: 203
Smältpunkt: 48…53 °C
Flampunkt: 113 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000249
CAS nummer: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8 II
Order Now
Specifikation testresultat
Appearance White to beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96.0 %