6-Bromo-1-hexene 97%
Leverantör: Thermo Fisher Scientific
Varning
364700010 364700050 364700250
ACRO364700010EA 640 SEK
ACRO364700010 ACRO364700050 ACRO364700250
6-Bromo-1-hexene 97%
6-Bromo-1-hexene
Formel: C₆H₁₁Br
Kokpunkt: 47…51 °C (16 mmHg)
Densitet: 1,217 g/cm³
Flampunkt: 52 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000269
CAS nummer: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %