2,3-Dibromopropene 80% tech.
Leverantör: Thermo Fisher Scientific
Fara
147740100 147740500 147742500
ACRO147740100EA 705 SEK
ACRO147740100 ACRO147740500 ACRO147742500
2,3-Dibromopropene 80% tech.
2,3-Dibromopropene
Formel: C₃H₄Br₂
Kokpunkt: 42…44 °C (17 mmHg)
Densitet: 2,13 g/cm³
Flampunkt: 81 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000211
CAS nummer: 513-31-5
EINECS: 208-155-8
UN: 3334
ADR: 9,III
Merck Index: 13,03050
Order Now
Specifikation testresultat
Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥75 %
Elemental Analysis 14.58 to 18.56 % (N)